6-Hydroxyindole 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol

OR7612-1G OR7612-250MG OR7612-5G
77411-254EA 57.06 USD
77411-254 77411-256 77411-258
6-Hydroxyindole 95%
6-Hydroxyindole
Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152101
CAS Number: 2380-86-1
EINECS: 417-020-4
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR