5-Aminopentan-1-ol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Amino-1-pentanol

OR51934-5G OR51934-100G OR51934-25G
77410-182EA 41.78 USD
77410-182 77410-178 77410-180
5-Aminopentan-1-ol 97%
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 120…122 °C/15.75 mmHg
Melting Pt: 33…35 °C
Density: 0.949 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
EINECS: 219-718-2
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR