3-Aminopropan-1-ol 99%

Supplier: APOLLO SCIENTIFIC

OR50998-100G OR50998-500G
77410-138EA 55.02 USD
77410-138 77410-140
3-Aminopropan-1-ol 99%
3-Aminopropan-1-ol
Formula: C₃H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00008223
CAS Number: 156-87-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR