3,3-Dimethylbutyric acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3,3-Dimethylbutanoic acid, tert-Butylacetic acid

OR0494-100G OR0494-500G
77402-100EA 24.46 USD
77402-100 77402-102
3,3-Dimethylbutyric acid
3,3-Dimethylbutyric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 185…190 °C
Melting Pt: –11 °C
Density: 0.9124 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002715
CAS Number: 1070-83-3
EINECS: 213-982-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR