2-Chloropyrimidine

Supplier: APOLLO SCIENTIFIC
Danger

OR0970-100G OR0970-25G OR0970-500G
77402-346EA 41.78 USD
77402-346 77402-348 77402-350
2-Chloropyrimidine
2-Chloropyrimidine
Formula: C₄H₃ClN₂
Boiling Pt: 75…76 °C/10 mmHg
Melting Pt: 62…67 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006060
CAS Number: 1722-12-9
EINECS: 217-020-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR