2,5-Difluorobenzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2724-100G PC2724-25G PC2724-5G
77413-776EA 130.88 USD
77413-776 77413-778 77413-780
2,5-Difluorobenzoyl chloride 98%
2,5-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 92…93 °C/34 mmHg
Density: 1.425 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00009929
CAS Number: 35730-09-7
EINECS: 252-701-8
UN: 2920
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR