2,2-Difluoroethanol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2805-100G PC2805-25G
77413-870EA 49.92 USD
77413-870 77413-872
2,2-Difluoroethanol 97%
2,2-Difluoroethanol

Can generate monofluoro acyl anion equivalent: Synlett, 503 (1994).

Formula: C₂H₄F₂O
Boiling Pt: 95…96 °C
Melting Pt: –28 °C
Density: 1.296 g/cm³ (25 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00042248
CAS Number: 359-13-7
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR