2,2,2-Tribromoethanol 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1882-5G
OR1882-100G
OR1882-25G
77406-416EA
61.12
USD
77406-416
77406-410
77406-412
2,2,2-Tribromoethanol 97%
2,2,2-Tribromoethanol
Formula:
Br₃CCH₂OH Boiling Pt: 92…93 °C/10 mmHg Melting Pt: 73…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004671 CAS Number: 75-80-9 EINECS: 200-903-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...