4-(Trifluoromethyl)anisole

Supplier: APOLLO SCIENTIFIC
Warning

PC8229-1G PC8229-25G PC8229-5G
77418-366EA 21.26 USD
77418-366 77418-368 77418-370
4-(Trifluoromethyl)anisole
4-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 166…170 °C
Melting Pt: –9 °C
Density: 1.2445 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD03094456
CAS Number: 402-52-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR