3-Fluorophenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC10393-25G PC10393-100G PC10393-10G
77412-814EA 38.72 USD
77412-814 77412-810 77412-812
3-Fluorophenyl isothiocyanate
3-Fluorophenyl isothiocyanate
Formula: C₇H₄FNS
Boiling Pt: 227 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004804
CAS Number: 404-72-8
EINECS: 206-967-7
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR