3-Fluorophenyl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4165-25G PC4165-5G
77415-178EA 72.34 USD
77415-178 77415-180
3-Fluorophenyl isocyanate 97%
3-Fluorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄FNO
Boiling Pt: 56…58 °C/14 mmHg
Density: 1.201 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002015
CAS Number: 404-71-7
EINECS: 206-966-1
UN: 3080
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR