(2-Methoxyethoxy)methyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride

OR4841-100G OR4841-25G
77410-054EA 200.7 USD
77410-054 77410-056
(2-Methoxyethoxy)methyl chloride, Technical Grade
(2-Methoxyethoxy)methyl chloride
Formula: ClCH₂OCH₂CH₂OCH₃
Boiling Pt: 50…52 °C/13 mmHg
Density: 1.091 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000888
CAS Number: 3970-21-6
EINECS: 223-589-8
UN: 2929
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR