4-Bromo-o-toluidine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-5-bromotoluene, 4-Bromo-2-methylaniline

OR21308-100G OR21308-25G OR21308-500G
77406-662EA 39.74 USD
77406-662 77406-664 77406-666
4-Bromo-o-toluidine
4-Bromo-o-toluidine
Formula: C₇H₈BrN
Boiling Pt: 240 °C
Melting Pt: 56…60 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007825
CAS Number: 583-75-5
UN: 3335
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR