4-(Difluoromethoxy)aniline 96%

Supplier: APOLLO SCIENTIFIC
Warning

PC2846E-100G PC2846E-25G PC2846E-5G
77413-912EA 125.32 USD
77413-912 77413-914 77413-916
4-(Difluoromethoxy)aniline 96%
4-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 231 °C
Density: 1.286 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085005
CAS Number: 22236-10-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR