2-Chloro-4-(trifluoromethyl)aniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-3-chloro-α,α,α-trifluorotoluene, 2-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-chlorobenzotrifluoride

PC1065-100G PC1065-25G
77412-944EA 58.08 USD
77412-944 77412-946
2-Chloro-4-(trifluoromethyl)aniline 97%
2-Chloro-4-(trifluoromethyl)aniline
Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
Boiling Pt: 214…218 °C
Density: 1.434 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042563
CAS Number: 39885-50-2
EINECS: 254-674-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR