α-Bromo-3-(trifluoromethoxy)toluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7438U-100G PC7438U-25G PC7438U-5G
77417-926EA 371.86 USD
77417-926 77417-928 77417-930
α-Bromo-3-(trifluoromethoxy)toluene 97%
α-Bromo-3-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Boiling Pt: 178…180 °C
Density: 1.572 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00061271
CAS Number: 159689-88-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR