2,4,6-Trifluorobenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-2,4,6-trifluorotoluene, 2-(Bromomethyl)-1,3,5-trifluorobenzene

PC7285T-1G PC7285T-250MG PC7285T-5G
77417-744EA 84.56 USD
77417-744 77417-746 77417-748
2,4,6-Trifluorobenzyl bromide 97%
2,4,6-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061210
CAS Number: 151411-98-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR