4-Methoxybenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

OR5028-100G OR5028-25G OR5028-500G
77410-104EA 63.16 USD
77410-104 77410-106 77410-108
4-Methoxybenzylamine 97%
4-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 64…66 °C
Density: 1.053 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008122
CAS Number: 2393-23-9
EINECS: 219-247-2
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR