3-Methoxybenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole

OR8643-100G OR8643-25G OR8643-5G
77411-424EA 186.44 USD
77411-424 77411-426 77411-428
3-Methoxybenzylamine 98%
3-Methoxybenzylamine
Formula: C₈H₁₁NO
Boiling Pt: 230…231 °C
Density: 1.7 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR