1,3-Difluoro-2-propanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC2878-25G PC2878-5G
77413-962EA 241.46 USD
77413-962 77413-964
1,3-Difluoro-2-propanol 97%
1,3-Difluoro-2-propanol
Formula: C₃H₆F₂O
Boiling Pt: 127 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00000452
CAS Number: 453-13-4
EINECS: 207-216-6
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR