α,α,α,3,4-Pentafluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC0225-100G PC0225-25G PC0225-5G
77412-462EA 142.32 USD
77412-462 77412-464 77412-466
α,α,α,3,4-Pentafluorotoluene 98%
α,α,α,3,4-Pentafluorotoluene
Formula: C₇H₃F₅
Boiling Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00077510
CAS Number: 32137-19-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR