(S)-(+)-1-Aminopropan-2-ol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR30797-1G OR30797-25G OR30797-5G
77408-956EA 21.26 USD
77408-956 77408-958 77408-960
(S)-(+)-1-Aminopropan-2-ol 98%
(S)-(+)-1-Aminopropan-2-ol
Formula: C₃H₉NO
Boiling Pt: 160 °C
Melting Pt: 19…26 °C
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064429
CAS Number: 2799-17-9
EINECS: 220-533-4
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR