4-Iodophenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Warning

OR401069-1G OR401069-5G
77409-506EA 32.6 USD
77409-506 77409-508
4-Iodophenyl isocyanate
4-Iodophenyl isocyanate

When melted, may contain small amount of sediment.

Formula: C₇H₄INO
Melting Pt: 44…48 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013875
CAS Number: 15845-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR