3-(Trifluoromethoxy)benzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7438O-1G PC7438O-25G PC7438O-5G
77417-918EA 21.26 USD
77417-918 77417-920 77417-922
3-(Trifluoromethoxy)benzylamine 97%
3-(Trifluoromethoxy)benzylamine
Formula: C₈H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00061267
CAS Number: 93071-75-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR