2,3,4-Trifluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7283D-100G PC7283D-25G PC7283D-5G
77417-708EA 69 USD
77417-708 77417-710 77417-712
2,3,4-Trifluorobenzonitrile 98%
2,3,4-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 116…118 °C/100 mmHg
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00013288
CAS Number: 143879-80-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR