Mercury(II) fluoride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5045-1G PC5045-5G
77416-374EA 21.26 USD
77416-374 77416-376
Mercury(II) fluoride 97%
Mercury(II) fluoride
Formula: F₂Hg
Melting Pt: 645 °C
Density: 8.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016139
CAS Number: 7783-39-3
UN: 2025
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR