2,6-Dichloro-4-(trifluoromethyl)aniline 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC2554-250G PC2554-25G PC2554-50G
77413-684EA 96.78 USD
77413-684 77413-686 77413-688
2,6-Dichloro-4-(trifluoromethyl)aniline 99%
2,6-Dichloro-4-(trifluoromethyl)aniline
Formula: C₇H₄Cl₂F₃N
Boiling Pt: 60…62 °C/1 mmHg
Melting Pt: 34…36 °C
Density: 1.532 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052918
CAS Number: 24279-39-8
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR