2-Chloro-4-fluoropyridine 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC5988-5G
PC5988-100G
PC5988-25G
77417-234EA
21.26
USD
77417-234
77417-230
77417-232
2-Chloro-4-fluoropyridine 98%
2-Chloro-4-fluoropyridine
Formula:
C₅H₃ClFN Boiling Pt: 140 °C Density: 1.456 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039345 CAS Number: 34941-91-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...