3-Chloropropionic acid 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Chloropropanoic acid

OR923420-100G OR923420-500G
77411-854EA 49.92 USD
77411-854 77411-856
3-Chloropropionic acid 98%
3-Chloropropionic acid
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002764
CAS Number: 107-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR