Erbium trichloride hexahydrate 99.99%

Supplier: APOLLO SCIENTIFIC
Warning

IN1696-100G IN1696-25G
77401-436EA 254.7 USD
77401-436 77401-438
Erbium trichloride hexahydrate 99.99%
Erbium trichloride hexahydrate
Formula: Cl₃Er·6H₂O
MW: 381.71
Melting Pt: 168
Storage Temperature: Ambient
MDL Number: MFCD00149693
CAS Number: 10025-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR