α-Bromo-2,3,4,5-tetrafluorotoluene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,3,4,5-Tetrafluorobenzyl bromide

PC0500-25G PC0500-5G
77412-584EA 429.94 USD
77412-584 77412-586
α-Bromo-2,3,4,5-tetrafluorotoluene
α-Bromo-2,3,4,5-tetrafluorotoluene
Formula: C₇H₃BrF₄
Boiling Pt: 107…110 °C/65 mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631453
CAS Number: 53001-71-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR