7-Bromoisatin 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione

OR14358-100G OR14358-25G OR14358-5G
77404-926EA 344.36 USD
77404-926 77404-928 77404-930
7-Bromoisatin 95%
7-Bromoisatin
Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 191…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00774354
CAS Number: 20780-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR