2,6-Dibromo-4-chloroaniline 97%

Supplier: APOLLO SCIENTIFIC

OR26854-100G OR26854-25G
77407-866EA 85.58 USD
77407-866 77407-868
2,6-Dibromo-4-chloroaniline 97%
2,6-Dibromo-4-chloroaniline
Formula: C₆H₄Br₂ClN
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00051750
CAS Number: 874-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR