1,3,5-Trimethyl-1H-pyrazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole

OR9220-5G OR9220-1G OR9220-25G
77411-774EA 50.94 USD
77411-774 77411-770 77411-772
1,3,5-Trimethyl-1H-pyrazole 98%
1,3,5-Trimethyl-1H-pyrazole
Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 170 °C
Melting Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00015536
CAS Number: 1072-91-9
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR