1-Phenylpiperidine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: N-Phenylpiperidine

OR909425-5G OR909425-1G OR909425-25G
77411-594EA 75.4 USD
77411-594 77411-590 77411-592
1-Phenylpiperidine 98%
1-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129777
CAS Number: 4096-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR