Benzenesulfonyl chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR16621-100G OR16621-250G OR16621-500G
77405-780EA 21.32 USD
77405-780 77405-782 77405-784
Benzenesulfonyl chloride 99%
Benzenesulphonyl chloride
Formula: C₆H₅ClO₂S
Boiling Pt: 251…252 °C
Melting Pt: 13…15 °C
Density: 1.384 g/cm³ (25 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00007426
CAS Number: 98-09-9
EINECS: 202-636-6
UN: 2225
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR