Methanesulphonic anhydride 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR17123-100G OR17123-500G
77405-962EA 47.8 USD
77405-962 77405-964
Methanesulphonic anhydride 98%
Methanesulphonic anhydride
Formula: C₂H₆O₅S₂
Boiling Pt: 125 °C/4 mmHg
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00007556
CAS Number: 7143-01-3
EINECS: 230-442-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR