2,3-Dimethylphenyl isocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR33664-1G OR33664-25G OR33664-5G
77409-210EA 22.26 USD
77409-210 77409-212 77409-214
2,3-Dimethylphenyl isocyanate 97%
2,3-Dimethylphenyl isocyanate

  • May form small amount of sediment.

Formula: C₉H₉NO
Boiling Pt: 88…90 °C/9.5 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00013851
CAS Number: 1591-99-7
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR