2-Iodophenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Danger

OR401067-1G OR401067-25G OR401067-5G
77409-494EA 55.02 USD
77409-494 77409-496 77409-498
2-Iodophenyl isocyanate
2-Iodophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄INO
Boiling Pt: 245 °C
Density: 1.891 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00673044
CAS Number: 128255-31-2
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR