2-(Trifluoromethoxy)benzylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2-(Trifluoromethoxy)phenyl)methanamine

PC7438NP-1G PC7438NP-25G PC7438NP-5G
77417-912EA 21.36 USD
77417-912 77417-914 77417-916
2-(Trifluoromethoxy)benzylamine 95%
2-(Trifluoromethoxy)benzylamine
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 77 °C/12 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00085176
CAS Number: 175205-64-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR