(Diethylamino)sulphur trifluoride 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
DAST
PC2563-100G
PC2563-25G
PC2563-5G
77413-696EA
199.7
USD
77413-696
77413-698
77413-700
(Diethylamino)sulphur trifluoride 95%
(Diethylamino)sulphur trifluoride
Versatile fluorinating agent: JOC., 1975, 40, 574.
Precaución: Has been reported to undergo explosive decomposition when heated above 90 °C.
Formula:
C₄H₁₀F₃NS MW: 161.19 g/mol Boiling Pt: 30…32 °C/3 mmHg Density: 1.22 g/cm³ (25 °C) Flash Pt: 23 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000363 CAS Number: 38078-09-0 EINECS: 253-771-2 UN: 2920 ADR: 8+3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...