2,2-Difluoro-1,3-benzodioxole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole

PC6674-100G PC6674-25G PC6674-500G
77417-478EA 56.2 USD
77417-478 77417-480 77417-482
2,2-Difluoro-1,3-benzodioxole 98%
2,2-Difluoro-1,3-benzodioxole
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Boiling Pt: 129…130 °C
Density: 1.308 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236217
CAS Number: 1583-59-1
EINECS: 216-431-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR