Antimony(III) fluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1140-1KG PC1140-250G PC1140-50G
77413-056EA 330.1 USD
77413-056 77413-058 77413-060
Antimony(III) fluoride 98%
Antimony(III) fluoride

A mild fluorinating agent; Fluorination activity may be increased by addition of Br2, Cl2, or SbCl 5. Swarts, Bull. Acad. Roy. Belg., 1892, 24, 309.

Formula: F₃Sb
Boiling Pt: 319 °C
Melting Pt: 292 °C
Density: 4.38 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR