2-Butynoic acid ethyl ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 2-butynoate

OR14510-1G OR14510-25G OR14510-5G
77404-992EA 21.28 USD
77404-992 77404-994 77404-996
2-Butynoic acid ethyl ester
2-Butynoic acid ethyl ester
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 164 °C
Density: 0.97 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00015182
CAS Number: 4341-76-8
EINECS: 224-395-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR