9,10-Phenanthrenequinone 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR322078-100G OR322078-25G OR322078-5G
77409-142EA 63.16 USD
77409-142 77409-144 77409-146
9,10-Phenanthrenequinone 98%
9,10-Phenanthrenequinone
Formula: C₁₄H₈O₂
Melting Pt: 201…213 °C
Flash Pt: 241 °C
Storage Temperature: Ambient
MDL Number: MFCD00001163
CAS Number: 84-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR