Chlorodifluoroacetic anhydride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1750-25G PC1750-100G
77413-300EA 135.5 USD
77413-300 77413-298
Chlorodifluoroacetic anhydride 98%
Chlorodifluoroacetic anhydride

Reagent for derivatisation of amines, alcohols etc, for GC purposes.

Formula: C₄Cl₂F₄O₃
Boiling Pt: 96…97 °C
Density: 1.395 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000774
CAS Number: 2834-23-3
EINECS: 220-603-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR