3,6-Dichloropyridazine 96%

Supplier: APOLLO SCIENTIFIC
Danger

OR21358-100G OR21358-500G
77406-670EA 21.26 USD
77406-670 77406-672
3,6-Dichloropyridazine 96%
3,6-Dichloropyridazine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR