1-Bromo-2-methoxyethane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromoethyl methyl ether

OR11137-100G OR11137-500G
77403-738EA 75.4 USD
77403-738 77403-740
1-Bromo-2-methoxyethane 98%
1-Bromo-2-methoxyethane
Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 109…110 °C
Density: 1.478 g/cm³ (25 °C)
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR