α-Bromo-2-(difluoromethoxy)toluene, Technical Grade

Supplier: APOLLO SCIENTIFIC

Synonyms: o-(Difluoromethoxy)benzyl bromide, 1-(Bromomethyl)-2-(difluoromethoxy)benzene, 2-(Difluoromethoxy)benzyl bromide

PC48840-25G PC48840-5G PC48840-100G
77415-814EA 124.3 USD
77415-814 77415-816 77415-812
α-Bromo-2-(difluoromethoxy)toluene, Technical Grade
α-Bromo-2-(difluoromethoxy)toluene
Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR