2-Bromobenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR26841-100G OR26841-25G OR26841-5G
77407-856EA 171.16 USD
77407-856 77407-858 77407-860
2-Bromobenzoyl chloride
2-Bromobenzoyl chloride
Formula: C₇H₄BrClO
Boiling Pt: 245 °C
Melting Pt: 8…10 °C
Density: 1.679 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000655
CAS Number: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR